missing translation for 'onlineSavingsMsg'
Learn More

Glucose-6-phosphate dehydrogenase, Leuconostoc mesenteroides, Thermo Scientific Chemicals

Glucose-6-phosphate dehydrogenase, derived from Leuconostoc mesenteroides, CAS# 9001-40-5, is an enzyme that catalyzes the conversion of p-glucose-6-phosphate to 6-phosphogluconolactone.

$2051.29 - $2051.29

Chemical Identifiers

CAS 9001-40-5
MDL Number MFCD00081656
Synonym G6PD
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60117JPL
View Documents
Thermo Scientific Chemicals
J60117JPL
10 kU
Each for $2,051.29
Only null left
 
Description

Description

Glucose-6-phosphate dehydrogenase, Leuconostoc mesenteroides is used to convert p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+. It is used as an intermediate, chemical and research purpose and also used fo analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Glucose-6-phosphate dehydrogenase, derived from Leuconostoc mesenteroides, is a cytosolic enzyme used to convert p-glucose-6-phosphate to 6-phosphogluconolactone
  • This enzyme can use either NAD or NADP+ as an electron acceptor to promote this conversion

Application

  • Glucose-6-phosphate dehydrogenase belongs to the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells, including erythrocytes, by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH)
  • Glucose-6-phosphate dehydrogenase can be used to measure hexose in solution
  • It is used as an intermediate for chemical and research purposes
  • It is used as a model to test the effect of seed protein fractions on enzyme protection during dehydration
  • It has been used in assays for nicotinamide adenine dinucleotide and tissue pyridine nucleotides
Specifications

Chemical Identifiers

9001-40-5
MFCD00081656
G6PD

Specifications

9001-40-5
MFCD00081656
G6PD
Lyophilized Powder
Plastic bottle
10 kU
It is soluble in water and dilute buffer.
Glucose-6-phosphate dehydrogenase
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-602-6

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only