missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glucagon-Like Peptide-1 (7-36) amide, human
Description
Glucagon-Like Peptide-1 is used as a Effector in the hormonal control of insulin secretion. The peptide fragment is secreted from the lower small intestine. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Common Name | Preproglucagon (78-107) amide, human |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| Molecular Weight (g/mol) | 3297.70 |
| Product Type | Glucagon-Like Peptide-1 (7-36) amide, human |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | GLP-1 (7-36) amide, human |
RUO – Research Use Only