missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon-Like Peptide-1 (7-36) amide, human

Catalog No. AAJ66197EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66197EXD 0.5 mg
AAJ66197MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66197EXD Supplier Thermo Scientific Chemicals Supplier No. J66197EXD

For cellular and molecular biology applications

Glucagon-Like Peptide-1 is used as a Effector in the hormonal control of insulin secretion. The peptide fragment is secreted from the lower small intestine. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Common Name Preproglucagon (78-107) amide, human
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 3297.70
Product Type Glucagon-Like Peptide-1 (7-36) amide, human
Quantity 0.5 mg
Regulatory Status RUO
Sequence GLP-1 (7-36) amide, human

RUO – Research Use Only