missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon (1-29) trifluoroacetate salt, human

Catalog No. AAJ60827EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60827EXD 0.5 mg
AAJ60827MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60827EXD Supplier Thermo Scientific Chemicals Supplier No. J60827EXD

For cellular and molecular biology applications

A peptide hormone that plays a role in maintaining glucose homeostasis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Format Powder
For Use With (Application) A peptide hormone that plays a role in maintaining glucose homeostasis
Molecular Weight (g/mol) 3482.74
Product Type Glucagon (1-29), human
Quantity 0.5 mg
Regulatory Status RUO

RUO – Research Use Only