missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8935 Hi-Vis Ranger Hat

Catalog No. p-7163129
Change view
Click to view available options
Color:
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19820725 Lime
19820726 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19820725 Supplier Ergodyne™ Supplier No. 23261
Only null left

Class Headwear Hi-Vis Ranger Hat features a wide brim and vented mesh panels.

  • ANSI-compliant 150D oxford polyester
  • Mesh venting panels
  • 1 in. ANSI-compliant reflective

Specifications

Size 2X-Large/3X-Large
Material Polyester
Color Lime
Size (English) 24 to 25.5 in.
Quantity 24/Pk.
Construction 150D Oxford Polyester
For Use With (Application) Road Construction, Utility Crew, Survey Crew, Construction