missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8928 Class E Hi-Vis Insulated Bibs

Catalog No. 19820979
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820979 Small
19820980 Medium
19820981 Large
19820982 X-Large
19820983 2X-Large
19820984 3X-Large
19820985 4X-Large
19820986 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19820979 Supplier Ergodyne™ Supplier No. 25522
Only null left

GloWear™ 8928 Hi-Vis Insulated Bibs provide conspicuity, warmth, and durability for the toughest conditions.

  • ANSI Class E Complaint for low light conditions
  • Made with tough 300D polyester oxford to withstand wear and tear
  • PU transparent coating provides wind and water-resistant protection
  • Scuff-resistant knee patches for extra durability
  • Black accents in high-wear areas to hide dirt and grime
  • Adjustable straps and buckle release for comfort and fit
  • 1 zippered chest pocket and 2 side hand pockets for storage
  • Boot zippers for easy on and off without removing footwear

Specifications

Size Small
Material Polyester
Color Yellow
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction