missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8925 Class E Thermal Pants

Catalog No. 19820868
Change view
Click to view available options
Color:
Orange
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19820868 Orange Small
19820869 Orange Medium
19820870 Orange Large
19820871 Orange X-Large
19820872 Orange 2X-Large
19820873 - 3X-Large
19820874 - 4X-Large
19820875 Orange 5X-Large
19820876 Yellow Small
19820877 - Medium
19820878 Yellow Large
19820879 Yellow X-Large
19820880 Yellow 2X-Large
19820881 Yellow 3X-Large
19820882 Yellow 4X-Large
19820883 Yellow 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19820868 Supplier Ergodyne™ Supplier No. 24442

Class E Thermal Pants feature breathable 3M™ Thinsulate™ warmth and waterproof comfort.

  • Weatherproof 100% polyester outer shell
  • Breathable PU coating
  • 80 g 3M™ Thinsulate™ lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket

Specifications

Size Small
Material Polyester
Color Orange
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction