missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8915BK Class E Black Bottom Rain Pants

Catalog No. 19820944
Change view
Click to view available options
Size:
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820944 2X-Large
19820946 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19820944 Supplier Ergodyne™ Supplier No. 25426

Class E Black Bottom Rain Pants offer breathable, waterproof, comfort with black accents to hide dirt.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg

Specifications

Size 2X-Large
Color Yellow
Waist Style Elastic with drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction