missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8915 Class E Rain Pants

Catalog No. 19820867
Change view
Click to view available options
Color:
Orange
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19820867 Yellow 5X-Large
19820852 Orange Small
19820853 Orange Medium
19820854 Orange Large
19820855 Orange X-Large
19820856 Orange 2X-Large
19820857 Orange 3X-Large
19820858 Orange 4X-Large
19820859 Orange 5X-Large
19820860 Yellow Small
19820861 Yellow Medium
19820862 Yellow -
19820863 Yellow X-Large
19820864 Yellow 2X-Large
19820865 Yellow -
19820866 Yellow 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19820867 Supplier Ergodyne™ Supplier No. 24429

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GloWear™ 8915 Class E Rain Pants designed to provide breathable, waterproof comfort.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket

Specifications

Size 5X-Large
Material Polyurethane
Color Yellow
Waist Style Elastic with drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction