missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8910BK Class E Black Bottom Hi-Vis Pants

Catalog No. p-7163105
Change view
Click to view available options
Size:
Small, Medium
Large, X-Large
4X-Large, 5X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820756 Small, Medium
19820757 Large, X-Large
19820759 4X-Large, 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19820756 Supplier Ergodyne™ Supplier No. 23953
Only null left

Class E Black Bottom Mesh Pants feature black high wear areas to hide dirt and keep worker looking clean.

  • ANSI-compliant polyester mesh (3.3 oz.)
  • 2 in. ANSI-compliant reflective
  • Drawstring waist
  • Wide leg openings with hook and loop closures
  • One right leg cargo pocket
  • Black accents on bottom of leg high wear areas

Specifications

Size Small, Medium
Product Type Hi-vis Mesh Pants
Material Polyester
Color Yellow
No. of Pockets 1
For Use With (Application) Road Construction, Utility Crew