missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8388 4-in-1, Type R, Class 3/2, Thermal High Visibility Jacket Kit

Catalog No. 19820992
Change view
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19820992 Lime Medium
19820993 Lime Large
19820994 Lime X-Large
19820995 Lime 2X-Large
19820996 Lime 3X-Large
19820997 Lime 4X-Large
19820998 Lime 5X-Large
19820999 Orange Small
19821000 Orange Medium
19821003 Orange Large
19821004 Orange X-Large
19821005 Orange 2X-Large
19910100 Orange 4X-Large
19910101 Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19820992 Supplier Ergodyne™ Supplier No. 25533

GloWear™ 8388 4-in-1 Thermal High Visibility Jacket Kit includes an outershell jacket and a thermal jacket.

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating
  • Inner jacket with 100 g 3M™ Thinsulate™ lining
  • Inner jacket zips out and sleeves zip off to create Class 2 thermal vest
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Black panels hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • External zipper chest pocket
  • Cellphone chest pocket
  • Clear ID badge holder
  • Dual mic tabs for clipping radios
  • YKK #5 zippers

Specifications

Size Medium
Material Oxford Polyester (300D)
Color Lime
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction