missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8009 Class E Two-Tone Mesh Leg Gaiters

Catalog No. 19910176
Change view
Click to view available options
Color:
Orange
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910176 Orange
19910177 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910176 Supplier Ergodyne™ Supplier No. 29091

GloWear™ Hi-Vis Leg Gaiters increase day and night visibility while staying cool with ultralight, breathable polyester mesh.

  • Ultralight ANSI-compliant polyester mesh is breathable and elastic to fit a variety of sizes
  • Two 2 in. ANSI-compliant 360 degree reflective tape with contrasting colors for enhanced visibility
  • Drawstring closures on top and bottom for quick adjustment
  • Knee-to-ankle coverage
  • Extra leg room slips easily over boots

Specifications

Certifications/Compliance ANSI/ISEA 107 Class E compliance
Color Orange
Dimensions (L x W) 47 x 56 cm
For Use With (Application) Road Construction, Utility Crew, Survey Crew
Material Polyester