missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ GloWear™ 8008 Class E Hi-Vis Leg Gaiters
Description
- Weatherproof polyester shell with PU coating
- 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
- Drawstring closures on top and bottom
Specifications
Specifications
| Closure Type | Drawstring |
| Color | Orange |
| For Use With (Application) | Road Construction, Utility Crew, Survey Crew, Construction |
| Material | Polyester |