missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8008 Class E Hi-Vis Leg Gaiters

Catalog No. 19910175
Change view
Click to view available options
Color:
Orange
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910175 Yellow
19910174 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910175 Supplier Ergodyne™ Supplier No. 29083

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hi-Vis Leg Gaiters add day and night conspicuity while protecting pants and boots from dirty outdoor environments.

  • Weatherproof polyester shell with PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Drawstring closures on top and bottom

Specifications

Closure Type Drawstring
Color Yellow
For Use With (Application) Road Construction, Utility Crew, Survey Crew, Construction
Material Polyester