missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ GloWear™ 8006 Hi-Vis Neck Shade
Description
- ANSI-compliant polyester solid fabric (3.6 oz.)
- Reflective binding
- Hook and loop closures
- Attaches to hard hat suspension headbands
Specifications
Specifications
| Color | Orange |
| For Use With (Application) | Road Construction, Utility Crew, Survey Crew |
| Material | Polyester |