missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8004 Hi-Vis Seat Belt Cover

Catalog No. 19910168
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19910168 Orange
19910169 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19910168 Supplier Ergodyne™ Supplier No. 29041

GloWear™ 8004 Hi-Vis Seat Belt Cover enchances the visibility of the driver.

  • ANSI-compliant polyester solid (3.6 oz.)
  • 2 in. ANSI-compliant reflective material
  • Fits standard width seat belt harness

Specifications

Closure Type Hook and Loop
Color Orange
For Use With (Equipment) Seat Belt
Material Polyester