missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Full Finger Cut Resistant Glove Liners
GREENER_CHOICE

Catalog No. 19126900
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19126900 Small
19126901 Medium
19126902 Large
19126903 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19126900 Supplier PURUS™ Supplier No. GLFFSCR

Glove liners are constructed of 13-gauge white nylon with HPPE to provide ANSI A2 cut protection. Designed to be worn under latex, PVC, or nitrile gloves to wick away moisture and prevent possible hand injuries.

  • Constructed of 13-gauge white nylon and HPPE
  • Designed to wick away moisture and prevent possible hand injury
  • Ideal for use under latex, nitrile, or PVC disposable gloves
  • An effective underglove for all cleanroom requirements
  • Color-coded cuffs for easy size identification
  • Launderable for reuse

Specifications

Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon, HPPE
Size Small
Finger Style Full
For Use With (Equipment) Latex, Nitrile, or PVC Disposable Gloves
Glove Cuff Color Green
Glove Cuff Style Knit
Hem Color Yellow
Series GLFF-CR
Yarn Gauge 13
Show More Show Less