missing translation for 'onlineSavingsMsg'
Learn More

Poltex Glove Box Holder ST DFS Item

Holders are constructed of break-resistant PETG and can be mounted horizontally or vertically. Each glove box has its own individual compartment for easy replacement of empty boxes.

Supplier:  Poltex 3GBSTW

 View more versions of this product

Catalog No. 22000138

Only null left
Explore available promotions

Description

Description

  • Constructed of clear, break-resistant PETG
  • Each box has its own individual compartment for easy replacement of empty boxes
  • Holder can be positioned vertically or horizontally
  • Glove boxes are shown for demonstration purposes only
  • Available in magnet mount (powerful magnets included) or wall mount (screws/anchors included)
Specifications

Specifications

Glove Box Holder
9 x 3.95 x 16.5 in.
22.9 cm
10 cm
41.9 cm
PETG
3
Mounting Hardware
Clear
9 in.
3.95 in.
16.5 in.
Disposable Glove Boxes
Keyhole Slots
3 Boxes
3 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions