Learn More
Poltex Glove Box Holder with Shelf
Break-resistant PETG glove box holder that holds 3 boxes of gloves, and other items on 2 shelves.
$532.93 - $645.13
Specifications
| Product Type | Glove Box Holder |
|---|---|
| Color | Clear |
| Material | PETG |
| Capacity | 3 Boxes |
Description
- Choose from wall mount/countertop placement (screws/anchors included), magnet mount (powerful magnets included), or tape mount (VHB tape included).
Specifications
| Glove Box Holder | |
| Clear | |
| PETG | |
| 3 Boxes |