missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Glove Box Gloves, Sleeves and Hands

Catalog No. p-172672
Change view
Click to view available options
Material:
Butyl
CSM
Neoprene
Size:
10
8.5
9
9.75
Universal
Thickness (English):
15 mil
17 mil
Color:
Black
White
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size Thickness (English) Color
1620023 Butyl 8.5 15 mil Black
1620021 Butyl 9.75 15 mil Black
1620022 CSM 8.5 15 mil White
1620020 CSM 9.75 15 mil White
162002 Neoprene 9.75 15 mil Black
161014 Neoprene 9 17 mil Black
1620015 Neoprene 10 17 mil Black
162005 Neoprene Universal 15 mil Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 1620023 Supplier Labconco™ Supplier No. 5005501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glove box gloves, sleeves and hands are available in a variety of sizes, materials and thicknesses to suit your specific application. Choose from neoprene, latex, CSM or butyl.

Glove Box Gloves - One pair/pack, one-piece, seamless, with O-rings

  • Neoprene - Size 9.75
    • 17 mil thickness
  • CSM (Chlorosulfonated polyethylene rubber) - Available in sizes 8.5 or 9.75
    • Also known as Hypalon
    • Resistant to abrasion, ozone and oxidizing chemicals
    • 15 mil thickness
  • Butyl - Available in sizes 8.5 and 9.75
    • Provides high impermeability to water vapor, gases and toxic chemicals
    • Resistant to oxygenated solvents and most oxidizing chemicals
    • 15 mil thickness

Glove Box Sleeves

  • Neoprene Straight Sleeves
    • For use with Neoprene or Latex Hands
    • With O-rings and springs

Glove Box Hands - For use with neoprene straight or accordian sleeves

  • Neoprene available in size 7.5, 9 or 10
    • 17 mil thickness
    • Lightweight and embossed with firm hold finish
  • Latex available in size 7 or 9
    • 9 mil thickness
    • White, surgical-type

Specifications

Color Black
Hand Ambidextrous
Latex Free Yes
Material Butyl
Product Type Glove Box Gloves
Size 8.5
Sterility Non-sterile
Thickness (English) 15 mil
Manufacturing/QMS Audit Standards ISO 9001
Model 5005501