Learn More
Thermo Scientific Chemicals Glove Bag Clamp
Catalog No. AA42909KF
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
AA42909KF 1 Ea.
1 options
Catalog No. AA42909KF Supplier Thermo Scientific Chemicals Supplier No. 042909KF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Quick-closing clamp
  • For use with 17 x 17 in. polyethylene glove bags
Specifications
For Use With (Equipment) Polyethylene Glove Bags (17 x 17 in.)
Quantity 1 Ea.
RUO – Research Use Only