Learn More
Globe Scientific Sterile Rigid Inoculating Loops with Needles
Catalog No. p-3242620
Change view
Click to view available options
Volume:
1 μL
10 μL
Packaging Type:
10/Peel Pack
20/Peel Pack
Individually Wrapped
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Packaging Type
22170204 10 μL 20/Peel Pack
22170207 1 μL 20/Peel Pack
22170206 10 μL Individually Wrapped
22170209 1 μL Individually Wrapped
22170205 10 μL 10/Peel Pack
22170208 1 μL 10/Peel Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22170204 Supplier Globe Scientific Supplier No. 2815F
Only null left
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
  • Certificate of Calibration comes with each case
  • Gamma Sterilized
Specifications
Disposable Single-use
Volume 10 μL
Material Polystyrene
Sterility Sterile
Color Yellow
Product Type Inoculating Loop, Rigid
Includes Needle and calibration certificate
Quantity 20
Packaging Type 20/Peel Pack