missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Polypropylene Test-Tube Racks

Takes up less benchtop space to provide larger work area

$66.16 - $71.16

Specifications

Material Polypropylene
View More Specs

Products 2
Catalog Number Mfr. No. Positions Holds Price Quantity & Availability  
Catalog Number Mfr. No. Positions Holds Price Quantity & Availability  
21200271
View Documents Promotion Details
Globe Scientific
456105
12 Tubes (12 mm)
Each for $71.16
Only null left
 
21200272
View Documents Promotion Details
Globe Scientific
456106
10 Tubes (0.94 mm)
Each for $66.16
Only null left
 
Description

Description

  • Made of polypropylene
  • Available in 12-place (12 to 13mm) and 10-place (16 to 18mm) formats
  • Autoclavable at 121°C for 20 minutes
Specifications

Specifications

Polypropylene
Videos
SDS
Documents

Documents