missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Formalin Filled Tite-Rite™ Containers

Catalog No. 22170402
Change view
Click to view available options
Capacity (Metric):
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22170402 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22170402 Supplier Globe Scientific Supplier No. 6518FL
Only null left

With dual-threaded polyethylene screw caps to provide leak-resistant, air-tight closures

  • Translucent, polypropylene containers
  • Prefilled with 10% neutral buffered formalin
  • Ribbed edges on cap and bottom for easy gripping
  • Comes with warning label, expiration date, lot number and white area for patient information

Compliance

95kPa compliant

Specifications

Capacity (English) 0.67 oz.
Capacity (Metric) 20 mL