Learn More
Globe Scientific Cuvette Rotor for ACL Coagulation Analyzers
Catalog No. 1437770
Change view
Click to view available options
For Use With (Equipment):
ACL Clinical Analyzers
Catalog No. For Use With (Equipment)
1437770 ACL Clinical Analyzers
1 options
Catalog No. 1437770
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
  • This 20-place cuvette rotors are designed for use with the IL ACL® models: 100, 200, 300, 1000, 2000, 3000, 6000 & 7000.
  • The cuvette rotors are packaged 25 per sleeve, 4 packs per case.
  • Please note: Not for use on the ACL 9000 & 10000.
Specifications
Type Cuvette Rotor
For Use With (Equipment) ACL Clinical Analyzers