missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bolle Safety Globe Goggle
Description
- Lightly translucent frame.
- Panoramic vision.
- Easily recycled.
- User-friendly color code.
Specifications
Specifications
| Lens Coating | Platinum Lite |
| Lens Tint | Clear |
| Product Type | Safety Goggles |
| Model | GLOBEN12W |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | PVC |
| UV Protection | None |
| Frame Type | Indirect Vented with Foam |
| Show More |