Learn More
Envigo RMS Teklad Global 18% Protein Rodent Diet, Fixed Formula, Irradiated, 113 lbs
Catalog No. NC2458161
Encompass
The Teklad Global 18% Protein Rodent Diet 2918 is a specialized irradiated diet optimized for the gestation, lactation, and growth of laboratory rodents. It ensures precise dietary control, devoid of alfalfa meal and fish meal, minimizing chlorophyll and carcinogenic nitrosamines. Enhanced with soybean meal, it regulates isoflavones for studies concerned with phytoestrogen levels. This high-quality rodent diet supports metabolic and toxicological research while promoting study integrity.
Catalog No. NC2458161 Supplier Envigo RMS Supplier No. T.2918.CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Teklad Global 18% Protein Rodent Diet 2918 is a specialized irradiated diet optimized for the gestation, lactation, and growth of laboratory rodents. It ensures precise dietary control, devoid of alfalfa meal and fish meal, minimizing chlorophyll and carcinogenic nitrosamines. Enhanced with soybean meal, it regulates isoflavones for studies concerned with phytoestrogen levels. This high-quality rodent diet supports metabolic and toxicological research while promoting study integrity.