missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gliquidone Sulfonamide, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Molecular Formula | C20H22N2O5S |
| Linear Formula | C20H22N2O5S |
| Quantity | 25 mg |
| Synonym | 4-[2-(3,4-Dihydro-7-methoxy-4,4-dimethyl-1,3-dioxo-2(1 H)-isoquinolinyl)ethyl]benzenesulfonamide |
| Formula Weight | 402.46 |
| Grade | BP |