missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gliclazide Impurity F, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 1076198-18-9 |
| Flash Point | Not Applicable |
| Molecular Formula | C15H21N3O3S |
| Linear Formula | C15H21N3O3S |
| Quantity | 30 mg |
| Synonym | N-[[(Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl)amino]carbonyl]-2-methylbenzenesulfonamide; 1-(Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl)-3-[(2-methylphenyl)sulphonyl]urea |
| Formula Weight | 323.41 |
| Grade | BP |