missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ GLI-1 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 2735 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB600-600 |
| Gene Alias | GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI |
| Gene Symbol | GLI1 |
| Quantity | 0.1 mg |
| Research Category | Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |