Electron Microscopy Sciences Lapping Vehicle Formula B Lube 16 OZ
Catalog No. 50997379
A universal lapping vehicle compatible with both water and oil soluble compounds. This very versatile formulation allows you to change materials and compounds and still provides excellent lubrication.
Catalog No.
50997379
Supplier
Electron Microscopy SciencesSupplier No.
5037610
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A universal lapping vehicle compatible with both water and oil soluble compounds. This very versatile formulation allows you to change materials and compounds and still provides excellent lubrication.