missing translation for 'onlineSavingsMsg'
Learn More

GlcNPhth[346Ac]beta(1-3)Gal[246Bn]-beta-MP 98.0+%, TCI America™

Catalog No. G0299200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0299200MG 200 mg
G02991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0299200MG Supplier TCI America Supplier No. G0299200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1820575-44-7
Molecular Formula C54H55NO16
Molecular Weight (g/mol) 974.025
InChI Key QHFVNDYHDLXLJC-JKFMBOLMSA-N
Synonym 4-Methoxyphenyl 3-O-(3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranosyl)-2,4,6-tri-O-benzyl-beta-D-galactopyranoside
PubChem CID 91659147
IUPAC Name [(2R,3S,4R,5R,6S)-3,4-diacetyloxy-5-(1,3-dioxoisoindol-2-yl)-6-[(2S,3R,4S,5S,6R)-2-(4-methoxyphenoxy)-3,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-4-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(C(OC(C2OCC3=CC=CC=C3)OC4=CC=C(C=C4)OC)COCC5=CC=CC=C5)OCC6=CC=CC=C6)N7C(=O)C8=CC=CC=C8C7=O)OC(=O)C)OC(=O)C

Specifications

Melting Point 175°C
Color White
Quantity 200 mg
Formula Weight 974.03
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNPhth[346Ac]beta(1-3)Gal[246Bn]-beta-MP
TSCA No
Recommended Storage Refrigerator