missing translation for 'onlineSavingsMsg'
Learn More

GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc 98.0+%, TCI America™

Catalog No. G04232MG
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04232MG 2 mg
G04235MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04232MG Supplier TCI America Supplier No. G04232MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108964-40-5
Molecular Formula C22H38N2O15

Specifications

Color White
Quantity 2 mg
Formula Weight 570.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc
TSCA No
Recommended Storage Freezer