missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 5mm (0.2in) dia, type 2

Catalog No. AA37998DM
Change view
Click to view available options
Quantity:
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA37998DM 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA37998DM Supplier Thermo Scientific Chemicals Supplier No. 037998DM
Only null left

Widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices. | CAS: 7440-44-0

Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]

Specifications

Packaging Tube
Quantity 100 mm
Solubility Information Insoluble in almost all solvents.
Chemical Name or Material Glassy carbon rod
EINECSNumber 231-153-3
RTECSNumber FF5250100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only