missing translation for 'onlineSavingsMsg'
Learn More

Glassy Carbon Lid;Top Dia (mm), 37;Bottom Dia (mm), 23

Catalog No. AA39020KT
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Shrink-wrapped
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA39020KT 1 Ea. Shrink-wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39020KT Supplier Thermo Scientific Chemicals Supplier No. 039020KT

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Packaging Shrink-wrapped
Quantity 1 Ea.
Chemical Name or Material Glassy Carbon Lid;Top Dia (mm)
TSCA U
Recommended Storage Ambient temperatures

RUO – Research Use Only