Learn More
Thermo Scientific Chemicals Glassy Carbon Boat;Vol (ml), 3;Length (mm), 53;Width (mm), 16;Ht (mm),10
Offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids
Supplier:  Thermo Scientific Chemicals 039027KT
 View more versions of this product
Catalog No. AA39027KT
Only null left
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Resistance to all wet decomposition agents and abrasive wear
  • No memory effects or contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistance to thermal shock allows rapid heating and cooling times
  • Good electrical conductivity
  • Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal­ growing such as doped halogenide crystals
Specifications
Chemical Identifiers
Glassy Carbon Boat
Specifications
Glassy Carbon Boat
53 x 16 x 10 mm
53 mm
3 mL
10 mm
16 mm
Product Suggestions
Product Suggestions