missing translation for 'onlineSavingsMsg'
Learn More

SP Hotpack™ Glassware Washer Accessories, Bottle Basket

Catalog No. 15365018
Change view
Click to view available options
Holds:
Up to 16, 1 L Bottles
Up to 23, 500 mL Bottles
Up to 24, 250 mL Bottles
Up to 40, 100 mL Bottles
Up to 69, 50 mL Bottles
Up to 9, 2 L Bottles
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
15365018 Up to 69, 50 mL Bottles
15365005 Up to 23, 500 mL Bottles
15365016 Up to 24, 250 mL Bottles
15365017 Up to 40, 100 mL Bottles
15365023 Up to 9, 2 L Bottles
15365007 Up to 16, 1 L Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15365018 Supplier SP Hotpack™ Supplier No. 100519220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bottle basket accessory designed for use with SP Hotpack™ undercounter, freestanding, and vertical stacking glassware washers.

  • Accessories maximize space, improve efficiency, and protect specialty glassware
  • For use in standard open racks
  • Made of stainless steel for long life and durability

Specifications

Material Stainless Steel
Product Type Bottle Holder
Holds Up to 69, 50 mL Bottles
For Use With (Equipment) 50 mL Bottles