Learn More
Kova Plastics by Alltrista Glasstic™ Slide II
Four-chambered slide
$478.29
Specifications
| Type | Slide II |
|---|---|
| For Use With (Application) | Microscopy |
| Material | Plastic with the optical qualities of glass |
| No. of Wells | 4 |
| Quantity | 100 |
Description
Slide ll is great for accurate, reproducible, and efficient standardized microscopic analysis.
- Made from optically clear plastic
- Each Slide ll features four individual fully segregated specimen wells
- Available with or without integrated counting grids
- Slides that include integrated counting grids provide results that have high correlation with results obtained with precision hemocytometers
- Capillary action reduces the likelihood of pipetting errors
- Suspends sediments homogeneously throughout the specimen
- Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
- Reproducible fluid distribution throughout each chamber
- Chamber walls prevent specimen leakage and cross-contamination
Specifications
| Slide II | |
| Plastic with the optical qualities of glass | |
| 100 |
| Microscopy | |
| 4 |