Learn More
MilliporeSigma™ Supelco™ GlasSeal™ Capillary Column Connector, Fused Silica
Catalog No. p-7191366
Change view
Click to view available options
Packaging Quantity:
25/Pk.
5/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
111010865 5/Pk.
111020772 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111010865 Supplier MilliporeSigma™ Supelco™ Supplier No. 23627
Only null left
Quickly connect two pieces of fused silica tubing of the same or different diameters without the need for tools.
The leak-free connection is useful to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases). Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Specifications
For Use With (Application) Gas Chromatography (GC)
Packaging Quantity 5/Pk.
GlasSeal is a trademark of Sigma-Aldrich Co. LLC