Learn More
MilliporeSigma™ Supelco™ Glass Wool
Catalog No. 111020928
Change view
Click to view available options
Product Type:
GC Accessories and Adsorbents
Non-Treated
Phosphoric Acid Treated
Silanized
Weight (Metric):
10 g
50 g
100 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Weight (Metric)
111020928 Non-Treated 50 g
111020909 GC Accessories and Adsorbents 10 g
111008042 GC Accessories and Adsorbents 100 g
111020914 Phosphoric Acid Treated 50 g
111020786 Silanized 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111020928 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20384
Only null left
Often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column
  • Made up of short glass fibres
  • Non-flammable and non-carcinogenic
  • Widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes
  • Suitable as noise absorbing material in concert halls, studios, and movie theatres
  • Can be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds
Specifications
Product Type Non-Treated
For Use With (Application) Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds
Material Glass Fiber
Weight (Metric) 50 g