Learn More
MilliporeSigma™ Supelco™ Glass Wool
Description
- Made up of short glass fibres
- Non-flammable and non-carcinogenic
- Widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes
- Suitable as noise absorbing material in concert halls, studios, and movie theatres
- Can be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds
Specifications
Specifications
| Product Type | GC Accessories and Adsorbents |
| For Use With (Application) | Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds |
| Material | Glass Fiber |
| Weight (Metric) | 10 g |