Learn More
MilliporeSigma™ Supelco™ Glass Wool
Catalog No. 111020786
Change view
Click to view available options
Product Type:
GC Accessories and Adsorbents
Non-Treated
Phosphoric Acid Treated
Silanized
Weight (Metric):
10 g
50 g
100 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Weight (Metric)
111020786 Silanized 250 g
111020909 GC Accessories and Adsorbents 10 g
111020928 Non-Treated 50 g
111008042 GC Accessories and Adsorbents 100 g
111020914 Phosphoric Acid Treated 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111020786 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20410
Only null left
Often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column
  • Made up of short glass fibres
  • Non-flammable and non-carcinogenic
  • Widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes
  • Suitable as noise absorbing material in concert halls, studios, and movie theatres
  • Can be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds
Specifications
Product Type Silanized
For Use With (Application) Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds
Material Glass Fiber
Weight (Metric) 250 g