missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Thistle Funnel, Stout Walled

Catalog No. S41696
Change view
Click to view available options
Height (Metric):
5.08 cm
10.16 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
S41696 10.16 cm
S41698 5.08 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S41696 Supplier Eisco™ Supplier No. CH0489A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glass thistle funnels are made from high quality borosilicate glass. Stout wall design. Autoclavable.

  • Made from high quality borosilicate glass

  • Stout wall design

  • Autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Diameter (Metric)Top 38 mm
Material Borosilicate Glass
Height (Metric) 10.16 cm
Autoclavable Autoclavable
Color Clear
Graduated No
For Use With (Application) transferring liquids
Quantity 1
Weight (Metric) 0.11
Height (English) 4
Product Type Stout Walled
Show More Show Less