missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Stirring Rods, 10 Pack
Glass stirring rods are made from high quality borosilicate glass and feature one spade and one button end.
Supplier: Eisco™ CH0755A
Description
Warranty
1 year limited manufacturer warranty
Chemical Identifiers
| Autoclavable | |
| 7 mm | |
| 10 Pk. | |
| Stirring Rod |
| Clear | |
| Borosilicate Glass | |
| Sphere | |
| Stirring |
Specifications
| 10 Pk. | |
| Autoclavable | |
| Borosilicate Glass | |
| Stirring | |
| 20 cm |
| Stirring Rod | |
| Sphere | |
| Clear | |
| 7 mm |