Learn More
Eisco™ Glass Rectangular Block
Catalog No. S14036
Change view
Click to view available options
Dimensions (L x W x H):
75 x 50 x 12 mm
75 x 50 x 18 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S14036 75 x 50 x 18 mm
S14035 75 x 50 x 12 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S14036 Supplier Eisco™ Supplier No. PH0566B
All angles accurately worked with edges slightly bevelled.
  • Made from imported optical glass
  • With optically worked surfaces
  • Glass free from bubbles
Specifications
Product Type Glass Rectangular Block
Description optically worked surfaces
Lens Material Imported Optical Glass
Dimensions (L x W x H) 75 x 50 x 18 mm