Learn More
Eisco™ Glass Rectangular Block
Description
- Made from imported optical glass
- With optically worked surfaces
- Glass free from bubbles
Specifications
Specifications
| Product Type | Glass Rectangular Block |
| Description | optically worked surfaces |
| Lens Material | Imported Optical Glass |
| Dimensions (L x W x H) | 75 x 50 x 12 mm |