Learn More
Corning™ PYREX™ Glass Pestle Tissue Grinders
Designed to handle both initial grinding and final homogenization
Supplier:  Corning™ 77245
 View more versions of this product
Catalog No. 0841421B
Only null left
Description
Description
Designed to handle both initial grinding and final homogenization in the same sequence of oper­ations. The lower conical section is intended primarily for the initial grinding. To assist in this work the clearance between mortar and pestle can be adjusted at will by raising or lowering the pestle. The upper cylindrical section is for the final homogenization.
Specifications
Chemical Identifiers
Non-sterile
Tissue Grinder
Pourout
Borosilicate Glass
Designed to handle both initial grinding and final homogenization
Specifications
Non-sterile
0.43 in.
5 mL
Tissue Grinder
8.85 in.
5.9 in.
0.98 in.
25 mm
11 mm
0.16 oz.
Borosilicate Glass
Designed to handle both initial grinding and final homogenization
225 mm
150 mm
Pourout
0.16 oz., 5 mL
Product Suggestions
Product Suggestions