missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Liebig Condensers
Description
- With screw threads
- Made from high quality Borosilicate 3.3 Glass
- Screw threads provide additional lab safety
- Screw thread joints allow flasks to connect firmly with other glassware
- Autoclavable
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Cool and condense hot vapor |
| Type | Liebig Condenser |
| Width (Metric) | 4 cm |
| Height (Metric) | 30 cm |
| Length (Metric) | 4 cm |
| Quantity | 1 |
| Standard Taper Joint No | 19/26 NS |