Learn More
BUCHI Glass Funnel Set For Funnel Rack
Catalog No. 05405936
Change view
Click to view available options
Glass Funnel Set
Catalog No.
05405936 Glass Funnel Set
1 options
Catalog No. 05405936 Supplier BUCHI Supplier No. 11074939
Glass funnels for the Pure Essential Chromatography Fraction Funnel Rack allow for large-volume fraction collection into bottles or carboys.
  • Unique shape for a secure fit in Pure Essential rack
  • Durable thick-walled glass
  • Easy clean up
Specifications
Type Glass Funnel
For Use With (Equipment) Funnel Rack