missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Gas Tube

Catalog No. S89303
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S89303 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S89303 Supplier Eisco™ Supplier No. CH0498C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eudiometer tube, used for gas collection and measurement

  • Made from high quality borosilicate glass
  • Used for collection and measurement of gases
  • White etched graduations
  • 100 mL capacity, 0.2 mL graduations

Warranty

1 year limited manufacturer warranty

Specifications

Color Clear
Grade 7 to Undergraduate
Age 12 to 20
For Use With (Application) Measuring gas
Includes Glass tube
Material Borosilicate Glass
Quantity 1
Capacity (Metric) 100 mL
Class Size 25 Students
Product Type Chemistry Kit